Tisk…

7-Bromo-5-[(trifluoromethyl)thio]-1,3-benzoxazole 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Bromo-5-[(trifluoromethyl)mercapto]benzo[d]oxazole, 7-Bromo-5-[(trifluoromethyl)sulphanyl]-1,3-benzoxazole

PC49344-1G PC49344-250MG
APOSPC49344-1GEA 11580 CZK
APOSPC49344-1G APOSPC49344-250MG
7-Bromo-5-[(trifluoromethyl)thio]-1,3-benzoxazole 97%
7-Bromo-5-[(trifluoromethyl)thio]-1,3-benzoxazole
Vzorec: C₈H₃BrF₃NOS
MW: 298,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24369702
CAS číslo: 1432075-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR