Tisk…

5-Bromo-2,4-difluorobenzoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-4,6-difluorobenzoic acid

PC49326-1G PC49326-5G PC49326-25G
APOSPC49326-1GEA 622 CZK
APOSPC49326-1G APOSPC49326-5G APOSPC49326-25G
5-Bromo-2,4-difluorobenzoic acid 97%
5-Bromo-2,4-difluorobenzoic acid
Vzorec: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Bod tání: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD04115714
CAS číslo: 28314-83-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR