Tisk…

3-Bromo-2-fluorophenacyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC49325-1G PC49325-250MG PC49325-5G
APOSPC49325-1GEA 9490 CZK
APOSPC49325-1G APOSPC49325-250MG APOSPC49325-5G
3-Bromo-2-fluorophenacyl bromide 95%
3-Bromo-2-fluorophenacyl bromide
Vzorec: C₈H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD16140216
CAS číslo: 1204333-47-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR