Tisk…

6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole

Dodavatel: Apollo Scientific

Synonyms: 6-Nitro-2-(perfluoroethyl)-1H-benzimidazole

PC49317-1G PC49317-250MG PC49317-5G
APOSPC49317-1GEA 4570 CZK
APOSPC49317-1G APOSPC49317-250MG APOSPC49317-5G
6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole
6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole
Vzorec: C₉H₄F₅N₃O₂
MW: 281,139 g/mol
Bod tání: 140…141 °C
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD18074389
CAS číslo: 4229-01-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR