Tisk…

2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,4,5,5-Tetramethyl-2-[2-(trifluoromethyl)pyridin-4-yl]-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine

PC49307-1G PC49307-250MG
APOSPC49307-1GEA 3280 CZK
APOSPC49307-1G APOSPC49307-250MG
2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester
2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester
Vzorec: C₁₂H₁₅BF₃NO₂
MW: 273.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607678
CAS číslo: 1036990-42-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR