5-Bromo-2-(trifluoromethoxy)benzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2-trifluoromethoxyphenylboronic acid, (5-Bromo-2-(trifluoromethoxy)phenyl)boronic acid
PC49296-1G
PC49296-5G
PC49296-25G
APOSPC49296-1GEA
953
CZK
APOSPC49296-1G
APOSPC49296-5G
APOSPC49296-25G
5-Bromo-2-(trifluoromethoxy)benzeneboronic acid
5-Bromo-2-(trifluoromethoxy)benzeneboronic acid
Vzorec:
C₇H₅BBrF₃O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11504857 CAS číslo: 1072951-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...