Tisk…

4-Bromo-3-(trifluoromethyl)benzeneboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Bromo-3-(trifluoromethyl)phenyl)boronic acid, 5-Borono-2-bromobenzotrifluoride, 4-Bromo-3-(trifluoromethyl)phenylboronic acid

PC49294-1G PC49294-250MG PC49294-5G
APOSPC49294-1GEA 3940 CZK
APOSPC49294-1G APOSPC49294-250MG APOSPC49294-5G
4-Bromo-3-(trifluoromethyl)benzeneboronic acid
4-Bromo-3-(trifluoromethyl)benzeneboronic acid
Vzorec: C₇H₅BBrF₃O₂
MW: 268.824 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181652
CAS číslo: 1310383-25-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR