Tisk…

4-Bromo-2-(trifluoromethyl)benzeneboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2-(trifluoromethyl)phenylboronic acid, 2-Borono-5-bromobenzotrifluoride, (4-Bromo-2-(trifluoromethyl)phenyl)boronic acid

PC49292-1G PC49292-5G PC49292-25G
APOSPC49292-1GEA 1780 CZK
APOSPC49292-1G APOSPC49292-5G APOSPC49292-25G
4-Bromo-2-(trifluoromethyl)benzeneboronic acid
4-Bromo-2-(trifluoromethyl)benzeneboronic acid
Vzorec: C₇H₅BBrF₃O₂
MW: 268.824 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447612
CAS číslo: 1394346-22-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR