Tisk…

3-Bromo-2-(difluoromethyl)thiophene 95%

Dodavatel: Apollo Scientific
Varování

PC49290-1G PC49290-5G PC49290-250MG
APOSPC49290-1GEA 6220 CZK
APOSPC49290-1G APOSPC49290-5G APOSPC49290-250MG
3-Bromo-2-(difluoromethyl)thiophene 95%
3-Bromo-2-(difluoromethyl)thiophene
Vzorec: C₅H₃BrF₂S
MW: 213,043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22580723
CAS číslo: 1403667-53-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR