Tisk…

3,6-Difluoro-2-methoxyphenol 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3,6-Difluoro-2-hydroxyanisole, 3,6-Difluoroguaiacol

PC49285-1G PC49285-250MG PC49285-5G
APOSPC49285-1GEA 8200 CZK
APOSPC49285-1G APOSPC49285-250MG APOSPC49285-5G
3,6-Difluoro-2-methoxyphenol 95%
3,6-Difluoro-2-methoxyphenol
Vzorec: C₇H₆F₂O₂
MW: 160.118 g/mol
Bod varu: 113.5…114.5 °C (37 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD12827564
CAS číslo: 75626-22-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR