Tisk…

4-Bromo-3-fluorophenylacetylene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-4-ethynyl-2-fluorobenzene

PC49282-1G PC49282-250MG PC49282-5G
APOSPC49282-1GEA 6290 CZK
APOSPC49282-1G APOSPC49282-250MG APOSPC49282-5G
4-Bromo-3-fluorophenylacetylene 98%
4-Bromo-3-fluorophenylacetylene
Vzorec: C₈H₄BrF
MW: 199,02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22460480
CAS číslo: 1191063-07-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR