Tisk…
4-Bromo-2-(trifluoromethoxy)benzotrifluoride 95%
Dodavatel: Apollo Scientific
Synonyms: 5-Bromo-2-(trifluoromethyl)phenyl trifluoromethyl ether, 4-Bromo-2-(trifluoromethoxy)-1-(trifluoromethyl)benzene
PC49263-1G PC49263-250MG
APOSPC49263-1GEA 17990 CZK
APOSPC49263-1G APOSPC49263-250MG
4-Bromo-2-(trifluoromethoxy)benzotrifluoride 95%
4-Bromo-2-(trifluoromethoxy)benzotrifluoride
Vzorec: C₈H₃BrF₆O
MW: 309.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415279
CAS číslo: 1396503-79-9