Tisk…

4-Bromo-5-fluoro-1H-benzimidazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-5-fluoro-1H-benzo[d]imidazole

PC49244-1G PC49244-250MG
APOSPC49244-1GEA 9920 CZK
APOSPC49244-1G APOSPC49244-250MG
4-Bromo-5-fluoro-1H-benzimidazole
4-Bromo-5-fluoro-1H-benzimidazole
Vzorec: C₇H₄BrFN₂
MW: 215,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22378819
CAS číslo: 1360962-58-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR