Tisk…

5-Bromo-2-fluoro-3-nitroaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-5-bromo-2-fluoronitrobenzene

PC49220-1G PC49220-250MG
APOSPC49220-1GEA 31680 CZK
APOSPC49220-1G APOSPC49220-250MG
5-Bromo-2-fluoro-3-nitroaniline
5-Bromo-2-fluoro-3-nitroaniline
Vzorec: C₆H₄BrFN₂O₂
MW: 235,011 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19441791
CAS číslo: 1313588-93-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR