Methyl 3-amino-2,6-difluorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC49203-1G
PC49203-5G
PC49203-25G
APOSPC49203-1GEA
622
CZK
APOSPC49203-1G
APOSPC49203-5G
APOSPC49203-25G
Methyl 3-amino-2,6-difluorobenzoate
Methyl 3-amino-2,6-difluorobenzoate
Vzorec:
C₈H₇F₂NO₂ Bod tání: 23…24 °C Storage Temperature: Ambient |
MDL Number:
MFCD13195461 CAS číslo: 84832-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...