Tisk…

3-(Difluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-3-(difluoromethyl)benzene, 3-(Difluoromethyl)benzyl Bromide, α-Bromo-α',α'-difluoro-m-xylene, 3-(Bromomethyl)benzal fluoride

PC49200-5G PC49200-1G PC49200-25G
APOSPC49200-5GEA 12570 CZK
APOSPC49200-5G APOSPC49200-1G APOSPC49200-25G
3-(Difluoromethyl)benzyl bromide 97%
3-(Difluoromethyl)benzyl bromide
Vzorec: C₈H₇BrF₂
MW: 221.042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18791600
CAS číslo: 1263178-51-3
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR