Tisk…

7-Bromo-5-fluoroquinoxaline 98%

Dodavatel: Apollo Scientific

Synonyms: 7-Bromo-5-fluoro-1,4-benzodiazine

PC49185-100MG PC49185-1G PC49185-250MG
APOSPC49185-100MGEA 1120 CZK
APOSPC49185-100MG APOSPC49185-1G APOSPC49185-250MG
7-Bromo-5-fluoroquinoxaline 98%
7-Bromo-5-fluoroquinoxaline
Vzorec: C₈H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD19053255
CAS číslo: 1210048-05-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR