3-Bromo-2,4-difluoronitrobenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-1,3-difluoro-4-nitrobenzene
PC49157-1G
PC49157-5G
PC49157-25G
APOSPC49157-1GEA
953
CZK
APOSPC49157-1G
APOSPC49157-5G
APOSPC49157-25G
3-Bromo-2,4-difluoronitrobenzene 98%
3-Bromo-2,4-difluoronitrobenzene
Vzorec:
C₆H₂BrF₂NO₂ Bod tání: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD12545882 CAS číslo: 103977-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...