4-Bromo-3-(trifluoromethoxy)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC49116-1G
PC49116-250MG
PC49116-5G
APOSPC49116-1GEA
1990
CZK
APOSPC49116-1G
APOSPC49116-250MG
APOSPC49116-5G
4-Bromo-3-(trifluoromethoxy)aniline 95%
4-Bromo-3-(trifluoromethoxy)aniline
Vzorec:
C₇H₅BrF₃NO MW: 256.02 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11506046 CAS číslo: 116369-25-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...