Tisk…

5-Fluoro-2-iodo-4-nitrotoluene

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-5-iodo-4-methylnitrobenzene, 1-Fluoro-4-iodo-5-methyl-2-nitrobenzene

PC49091-1G PC49091-250MG PC49091-5G
APOSPC49091-1GEA 4800 CZK
APOSPC49091-1G APOSPC49091-250MG APOSPC49091-5G
5-Fluoro-2-iodo-4-nitrotoluene
5-Fluoro-2-iodo-4-nitrotoluene
Vzorec: C₇H₅FINO₂
MW: 281.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27937150
CAS číslo: 1803811-31-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR