Tisk…

3-Amino-4-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

PC49084-1G PC49084-250MG PC49084-5G
APOSPC49084-1GEA 705 CZK
APOSPC49084-1G APOSPC49084-250MG APOSPC49084-5G
3-Amino-4-(trifluoromethyl)benzonitrile
3-Amino-4-(trifluoromethyl)benzonitrile
Vzorec: C₈H₅F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD18072580
CAS číslo: 1220630-83-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR