Tisk…

4-Bromo-2-methoxybenzotrifluoride

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-(trifluoromethyl)anisole, 5-Bromo-2-(trifluoromethyl)phenyl methyl ether, 4-Bromo-2-methoxy-1-(trifluoromethyl)benzene

PC49083-1G PC49083-25G PC49083-5G
APOSPC49083-1GEA 910 CZK
APOSPC49083-1G APOSPC49083-25G APOSPC49083-5G
4-Bromo-2-methoxybenzotrifluoride
4-Bromo-2-methoxybenzotrifluoride
Vzorec: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194486
CAS číslo: 944805-63-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR