2'-Fluoro-3'-methoxyacetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Acetyl-2-fluoroanisole, 1-(2-Fluoro-3-methoxyphenyl)ethan-1-one
PC49075-1G
PC49075-250MG
PC49075-5G
APOSPC49075-1GEA
7670
CZK
APOSPC49075-1G
APOSPC49075-250MG
APOSPC49075-5G
2'-Fluoro-3'-methoxyacetophenone 97%
2'-Fluoro-3'-methoxyacetophenone
Vzorec:
C₉H₉FO₂ MW: 168,165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13194330 CAS číslo: 208777-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...