Tisk…

4-Bromo-7-fluoroindan-1-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2,3-dihydro-7-fluoro-1H-inden-1-one, 4-Bromo-7-fluoro-2,3-dihydro-1H-inden-1-one, 4-Bromo-2,3-dihydro-7-fluoro-1-oxo-1H-indene

PC49069-1G PC49069-250MG PC49069-5G
APOSPC49069-1GEA 1410 CZK
APOSPC49069-1G APOSPC49069-250MG APOSPC49069-5G
4-Bromo-7-fluoroindan-1-one
4-Bromo-7-fluoroindan-1-one
Vzorec: C₉H₆BrFO
MW: 229,046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17677340
CAS číslo: 1003048-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR