3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Methyl-7-(trifluoromethyl)-1H-indazol-3-amine
PC49029-1G
PC49029-250MG
APOSPC49029-1GEA
11580
CZK
APOSPC49029-1G
APOSPC49029-250MG
3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole
3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole
Vzorec:
C₉H₈F₃N₃ MW: 215.175 g/mol Bod tání: 95…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD11847095 CAS číslo: 1260678-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...