Tisk…

3-Bromo-4-fluoro-2-methoxyaniline

Dodavatel: Apollo Scientific
Varování

PC49008-1G PC49008-250MG
APOSPC49008-1GEA 7940 CZK
APOSPC49008-1G APOSPC49008-250MG
3-Bromo-4-fluoro-2-methoxyaniline
3-Bromo-4-fluoro-2-methoxyaniline
Vzorec: C₇H₇BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD18071064
CAS číslo: 1257535-12-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR