2-Amino-3-bromo-4-fluorobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC49007-1G
PC49007-250MG
APOSPC49007-1GEA
12570
CZK
APOSPC49007-1G
APOSPC49007-250MG
2-Amino-3-bromo-4-fluorobenzonitrile
2-Amino-3-bromo-4-fluorobenzonitrile
Vzorec:
C₇H₄BrFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD18071063 CAS číslo: 1093951-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...