Tisk…

1-Bromo-5-chloro-2-fluoro-3-iodobenzene 95%

Dodavatel: Apollo Scientific

PC48952-1G PC48952-250MG
APOSPC48952-1GEA 16340 CZK
APOSPC48952-1G APOSPC48952-250MG
1-Bromo-5-chloro-2-fluoro-3-iodobenzene 95%
1-Bromo-5-chloro-2-fluoro-3-iodobenzene
Vzorec: C₆H₂BrClFI
MW: 335,34 g/mol
Storage Temperature: Ambient
CAS číslo: 2135861-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR