Ethyl 2-{[(4-fluoroanilino)carbothioyl]amino}acetate
Dodavatel: Apollo Scientific
Synonyms:
1-(Ethoxycarbonylmethyl)-3-(4-fluorophenyl)-2-thiourea
PC4895-1G
PC4895-500MG
APOSPC4895-1GEA
16800
CZK
APOSPC4895-1G
APOSPC4895-500MG
Ethyl 2-{[(4-fluoroanilino)carbothioyl]amino}acetate
Ethyl 2-{[(4-fluoroanilino)carbothioyl]amino}acetate
Vzorec:
C₁₁H₁₃FN₂O₂S MW: 256.297 g/mol Bod tání: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00975556 CAS číslo: 338773-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...