Tisk…

5-Bromo-2,3,4-trifluorobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC48931-1G PC48931-250MG PC48931-5G
APOSPC48931-1GEA 4140 CZK
APOSPC48931-1G APOSPC48931-250MG APOSPC48931-5G
5-Bromo-2,3,4-trifluorobenzoic acid 95%
5-Bromo-2,3,4-trifluorobenzoic acid
Vzorec: C₇H₂BrF₃O₂
MW: 254,989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756771
CAS číslo: 212631-85-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR