Tisk…

4-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC48918-1G PC48918-250MG PC48918-5G
APOSPC48918-1GEA 3270 CZK
APOSPC48918-1G APOSPC48918-250MG APOSPC48918-5G
4-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
4-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Vzorec: C₇H₂BrClF₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD32070722
CAS číslo: 2366994-51-4
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR