Tisk…

3-Fluoro-4-nitrobenzotrifluoride 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Fluoro-4-(trifluoromethyl)nitrobenzene, 2-Fluoro-1-nitro-4-(trifluoromethyl)benzene

PC48831-1G PC48831-25G PC48831-5G
APOSPC48831-1GEA 827 CZK
APOSPC48831-1G APOSPC48831-25G APOSPC48831-5G
3-Fluoro-4-nitrobenzotrifluoride 95%
3-Fluoro-4-nitrobenzotrifluoride
Vzorec: C₇H₃F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD03840208
CAS číslo: 402-12-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR