5-Bromo-2,3,4-trifluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC48807-1G
PC48807-250MG
PC48807-5G
APOSPC48807-1GEA
1670
CZK
APOSPC48807-1G
APOSPC48807-250MG
APOSPC48807-5G
5-Bromo-2,3,4-trifluorobenzonitrile 95%
5-Bromo-2,3,4-trifluorobenzonitrile
Vzorec:
C₇HBrF₃N Storage Temperature: Ambient |
MDL Number:
MFCD26679060 CAS číslo: 1503508-74-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...