4-Bromo-2-fluorophenyl acetate
Dodavatel: Apollo Scientific
|
Varování
|
PC48806-1G
PC48806-250MG
PC48806-5G
APOSPC48806-1GEA
1670
CZK
APOSPC48806-1G
APOSPC48806-250MG
APOSPC48806-5G
4-Bromo-2-fluorophenyl acetate
4-Bromo-2-fluorophenyl acetate
Vzorec:
C₈H₆BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28156435 CAS číslo: 161424-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...