Tisk…

2-Bromo-6-(difluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC48802-1G PC48802-250MG PC48802-5G
APOSPC48802-1GEA 14980 CZK
APOSPC48802-1G APOSPC48802-250MG APOSPC48802-5G
2-Bromo-6-(difluoromethyl)benzoic acid 95%
2-Bromo-6-(difluoromethyl)benzoic acid
Vzorec: C₈H₅BrF₂O₂
MW: 251.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31635696
CAS číslo: 1782342-72-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR