Tisk…

1-Bromo-3-(difluoromethyl)-2-iodobenzene 95%

Dodavatel: Apollo Scientific
Varování

PC48801-1G PC48801-250MG PC48801-5G
APOSPC48801-1GEA 16470 CZK
APOSPC48801-1G APOSPC48801-250MG APOSPC48801-5G
1-Bromo-3-(difluoromethyl)-2-iodobenzene 95%
1-Bromo-3-(difluoromethyl)-2-iodobenzene
Vzorec: C₇H₄BrF₂I
Storage Temperature: Ambient
MDL Number: MFCD18391108
CAS číslo: 1261775-78-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR