Tisk…

2,4-Dichloro-6-fluoroanisole 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,5-Dichloro-3-fluoro-2-methoxybenzene

PC48795-1G PC48795-5G PC48795-25G
APOSPC48795-1GEA 8870 CZK
APOSPC48795-1G APOSPC48795-5G APOSPC48795-25G
2,4-Dichloro-6-fluoroanisole 99%
2,4-Dichloro-6-fluoroanisole
Vzorec: C₇H₅Cl₂FO
MW: 195,018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28785944
CAS číslo: 54300-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR