2-Bromo-5-(trifluoromethoxy)phenylacetylene 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-2-ethynyl-4-(trifluoromethoxy)benzene
PC48788-1G
PC48788-250MG
APOSPC48788-1GEA
3910
CZK
APOSPC48788-1G
APOSPC48788-250MG
2-Bromo-5-(trifluoromethoxy)phenylacetylene 95%
2-Bromo-5-(trifluoromethoxy)phenylacetylene
Vzorec:
C₉H₄BrF₃O MW: 265.027 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31726320 CAS číslo: 2379918-32-6 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...