Tisk…

3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-3-ethynyl-5-(trifluoromethoxy)benzene

PC48787-1G PC48787-250MG PC48787-5G
APOSPC48787-1GEA 3270 CZK
APOSPC48787-1G APOSPC48787-250MG APOSPC48787-5G
3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%
3-Bromo-5-(trifluoromethoxy)phenylacetylene
Vzorec: C₉H₄BrF₃O
MW: 265.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31726319
CAS číslo: 2366994-52-5
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR