Tisk…

2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one

Dodavatel: Apollo Scientific
Varování

PC48783-1G PC48783-250MG PC48783-5G
APOSPC48783-1GEA 4470 CZK
APOSPC48783-1G APOSPC48783-250MG APOSPC48783-5G
2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one
2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one
Vzorec: C₆H₅F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16988116
CAS číslo: 175354-56-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR