2-Fluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Dodavatel: Apollo Scientific
|
Varování
|
PC48782-1G
PC48782-250MG
APOSPC48782-1GEA
6220
CZK
APOSPC48782-1G
APOSPC48782-250MG
2-Fluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
2-Fluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Vzorec:
C₁₀H₉F₄NO₂ MW: 251,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27946933 CAS číslo: 198967-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...