Tisk…

2-Chloro-4-fluoro-5-iodonitrobenzene 96%

Dodavatel: Apollo Scientific
Varování

PC48773-1G PC48773-250MG PC48773-5G
APOSPC48773-1GEA 9330 CZK
APOSPC48773-1G APOSPC48773-250MG APOSPC48773-5G
2-Chloro-4-fluoro-5-iodonitrobenzene 96%
2-Chloro-4-fluoro-5-iodonitrobenzene
Vzorec: C₆H₂ClFINO₂
MW: 301.441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723635
CAS číslo: 2244086-60-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR