Tisk…

Perfluoro(1,3-dimethyl-5-ethylcyclohexane)

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,4-Bis(trifluoromethyl)-1,1,2,3,3,4,5,5,6-nonafluoro-6-(pentafluoroethyl)cyclohexane

PC4877-100G PC4877-25G PC4877-5G
APOSPC4877-100GEA 8500 CZK
APOSPC4877-100G APOSPC4877-25G APOSPC4877-5G
Perfluoro(1,3-dimethyl-5-ethylcyclohexane)
Perfluoro(1,3-dimethyl-5-ethylcyclohexane)
Vzorec: C₁₀F₂₀
MW: 500,075 g/mol
Bod varu: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD04972337
CAS číslo: 155609-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR