2,3,5,6-Tetrafluorobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC48765-1G
PC48765-250MG
APOSPC48765-1GEA
6720
CZK
APOSPC48765-1G
APOSPC48765-250MG
2,3,5,6-Tetrafluorobenzenesulphonyl chloride
2,3,5,6-Tetrafluorobenzenesulphonyl chloride
Vzorec:
C₆HClF₄O₂S MW: 248,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06409089 CAS číslo: 776-09-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...