Tisk…

2-Bromo-3-(2,2,2-trifluoroethoxy)pyridine 95%

Dodavatel: Apollo Scientific
Varování

PC48762-1G PC48762-250MG PC48762-5G
APOSPC48762-1GEA 14060 CZK
APOSPC48762-1G APOSPC48762-250MG APOSPC48762-5G
2-Bromo-3-(2,2,2-trifluoroethoxy)pyridine 95%
2-Bromo-3-(2,2,2-trifluoroethoxy)pyridine
Vzorec: C₇H₅BrF₃NO
MW: 256,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11878543
CAS číslo: 256473-06-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR