7-Fluorobenzo[b]thiophene-2-carboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC48755-1G
PC48755-250MG
PC48755-5G
APOSPC48755-1GEA
7250
CZK
APOSPC48755-1G
APOSPC48755-250MG
APOSPC48755-5G
7-Fluorobenzo[b]thiophene-2-carboxaldehyde
7-Fluorobenzo[b]thiophene-2-carboxaldehyde
Vzorec:
C₉H₅FOS Storage Temperature: Ambient |
MDL Number:
MFCD17169315 CAS číslo: 698367-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...