5-Bromo-2-chloro-4-fluoroaniline 95%
Dodavatel: Apollo Scientific
PC48742-1G
PC48742-5G
APOSPC48742-1GEA
2200
CZK
APOSPC48742-1G
APOSPC48742-5G
5-Bromo-2-chloro-4-fluoroaniline 95%
5-Bromo-2-chloro-4-fluoroaniline
Vzorec:
C₆H₄BrClFN MW: 224.458 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21603962 CAS číslo: 305795-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...