2-Chloro-4-fluoro-3-methylbenzaldehyde 98%
Dodavatel: Apollo Scientific
PC48741-1G
PC48741-250MG
APOSPC48741-1GEA
8500
CZK
APOSPC48741-1G
APOSPC48741-250MG
2-Chloro-4-fluoro-3-methylbenzaldehyde 98%
2-Chloro-4-fluoro-3-methylbenzaldehyde
Vzorec:
C₈H₆ClFO MW: 172,584 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15528861 CAS číslo: 1260764-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...