6-Bromo-2-fluoro-3-methoxybenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC48737-10G
PC48737-1G
PC48737-5G
APOSPC48737-10GEA
9920
CZK
APOSPC48737-10G
APOSPC48737-1G
APOSPC48737-5G
6-Bromo-2-fluoro-3-methoxybenzaldehyde 98%
6-Bromo-2-fluoro-3-methoxybenzaldehyde
Vzorec:
C₈H₆BrFO₂ MW: 233,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261251 CAS číslo: 853792-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...