Tisk…

6-Bromo-2-fluoro-3-iodobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

PC48735-1G PC48735-250MG
APOSPC48735-1GEA 18620 CZK
APOSPC48735-1G APOSPC48735-250MG
6-Bromo-2-fluoro-3-iodobenzonitrile 95%
6-Bromo-2-fluoro-3-iodobenzonitrile
Vzorec: C₇H₂BrFIN
MW: 325,904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31629889
CAS číslo: 217816-65-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR